Assay Details
Gene Symbol: RGD1564450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106377.1 | NP_001099847.1 | 1 - 2 | 286 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106377.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
