Assay Details
| Gene Symbol: Gstt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109675.1 | NP_001103145.1 | 3 - 4 | 448 | 103 | |
| GenBank mRNA | BC097423.1 | - | 3 - 4 | 432 | 7455039 | 103 |
More Information
Updated Mapping Information
Set Membership: |
|

Gstt4
Genomic Map