Assay Details
| Gene Symbol: Ube2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108530.1 | NP_001102000.1 | 5 - 6 | 396 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69650 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000480 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000480 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P000480 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P274550 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31188 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19967 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10829699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829700 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829701 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10829694 | - | - |
| Rat Genome 230 2.0 Array | 1390259_at | - | - |
| Rat Genome 230 2.0 Array | 1385548_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390259_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385548_at | - | - |
| Affymetrix HT Rat Focus Array | 1390259_at | - | - |
| Affymetrix HT Rat Focus Array | 1385548_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390259_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385548_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000004769.rn.1 | TC2000000507.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000004771.rn.1 | TC2000000507.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000004767.rn.1 | TC2000000507.rn.1 | RTA TC |
| Rat Expression Set 230 | 1385548_at | - | - |
| Rat Expression Set 230 | 1390259_PM_at | - | - |
| Rat Expression Set 230 | 1390259_at | - | - |
| Rat Expression Set 230 | 1385548_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10829694 | - | - |
| Rat Genome U34 Set | AA942808_at | - | - |
| Rat Genome U34 Set | rc_AI231279_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22196065 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351247 | - | - |

Ube2d1
Genomic Map