Assay Details
| Gene Symbol: LOC103694875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773833.1 | XP_008772055.1 | 1 - 2 | 1514 | 92 | |
| XM_008773837.1 | XP_008772059.1 | 1 - 2 | 1515 | 92 | ||
| XM_008773840.1 | XP_008772062.1 | 1 - 2 | 1510 | 92 | ||
| XM_008773841.1 | XP_008772063.1 | 1 - 2 | 1511 | 92 | ||
| Gene Symbol: Derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109577.1 | NP_001103047.2 | 1 - 2 | 122 | 92 | |
| XM_006256325.2 | XP_006256387.1 | 1 - 2 | 1511 | 92 | ||
| XM_008772918.1 | XP_008771140.1 | 1 - 2 | 1514 | 92 | ||
| XM_008772922.1 | XP_008771144.1 | 1 - 2 | 1515 | 92 | ||
| GenBank mRNA | BC167034.1 | - | 1 - 2 | 79 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103694875
Genomic Map