Assay Details
| Gene Symbol: LOC103694875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773829.1 | XP_008772051.1 | 2 - 3 | 544 | 140 | |
| XM_008773830.1 | XP_008772052.1 | 2 - 3 | 544 | 140 | ||
| XM_008773831.1 | XP_008772053.1 | 2 - 3 | 544 | 140 | ||
| XM_008773832.1 | XP_008772054.1 | 2 - 3 | 544 | 140 | ||
| XM_008773833.1 | XP_008772055.1 | 4 - 5 | 1741 | 140 | ||
| XM_008773834.1 | XP_008772056.1 | 2 - 3 | 544 | 140 | ||
| XM_008773835.1 | XP_008772057.1 | 4 - 5 | 1738 | 140 | ||
| XM_008773836.1 | XP_008772058.1 | 3 - 4 | 433 | 140 | ||
| XM_008773837.1 | XP_008772059.1 | 3 - 4 | 1676 | 140 | ||
| XM_008773838.1 | XP_008772060.1 | 2 - 3 | 544 | 140 | ||
| XM_008773839.1 | XP_008772061.1 | 2 - 3 | 544 | 140 | ||
| XM_008773840.1 | XP_008772062.1 | 4 - 5 | 1737 | 140 | ||
| XM_008773841.1 | XP_008772063.1 | 4 - 5 | 1738 | 140 | ||
| XM_008773842.1 | XP_008772064.1 | 4 - 5 | 1735 | 140 | ||
| Gene Symbol: Derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109577.1 | NP_001103047.2 | 4 - 5 | 349 | 140 | |
| XM_006256325.2 | XP_006256387.1 | 4 - 5 | 1738 | 140 | ||
| XM_006256326.2 | XP_006256388.1 | 4 - 5 | 1735 | 140 | ||
| XM_008772914.1 | XP_008771136.1 | 2 - 3 | 544 | 140 | ||
| XM_008772915.1 | XP_008771137.1 | 2 - 3 | 544 | 140 | ||
| XM_008772916.1 | XP_008771138.1 | 2 - 3 | 544 | 140 | ||
| XM_008772917.1 | XP_008771139.1 | 2 - 3 | 544 | 140 | ||
| XM_008772918.1 | XP_008771140.1 | 4 - 5 | 1741 | 140 | ||
| XM_008772919.1 | XP_008771141.1 | 2 - 3 | 544 | 140 | ||
| XM_008772920.1 | XP_008771142.1 | 4 - 5 | 1738 | 140 | ||
| XM_008772921.1 | XP_008771143.1 | 3 - 4 | 433 | 140 | ||
| XM_008772922.1 | XP_008771144.1 | 3 - 4 | 1676 | 140 | ||
| XM_008772923.1 | XP_008771145.1 | 2 - 3 | 544 | 140 | ||
| XM_008772924.1 | XP_008771146.1 | 2 - 3 | 544 | 140 | ||
| GenBank mRNA | BC167034.1 | - | 4 - 5 | 306 | 140 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103694875
Genomic Map