Assay Details
Gene Symbol: Specc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039455.1 | NP_001034544.1 | 15 - 16 | 3451 | 62 | |
XM_006256339.2 | XP_006256401.1 | 15 - 16 | 3433 | 62 | ||
GenBank mRNA | AY884296.1 | - | 15 - 16 | 3451 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P127802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117050 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P127802 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P127802 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P127802 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P127802 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 21184 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94276 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3391 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52753 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92452 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67063 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10832590 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832599 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832586 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832589 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832597 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832587 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832595 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832594 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832600 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832593 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832588 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832601 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832591 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832592 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832596 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832598 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10832584 | - | - |
Rat Genome 230 2.0 Array | 1380790_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383723_at | - | - |
Rat Genome 230 2.0 Array | 1383723_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373104_at | - | - |
Rat Genome 230 2.0 Array | 1380790_at | - | - |
Rat Genome 230 2.0 Array | 1373104_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380790_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380790_at | - | - |
Affymetrix HT Rat Focus Array | 1373104_at | - | - |
Affymetrix HT Rat Focus Array | 1373104_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2000011272.rn.1 | TC2000001365.rn.1 | RTA TC |
Rat Expression Set 230 | 1373104_PM_at | - | - |
Rat Expression Set 230 | 1373104_at | - | - |
Rat Expression Set 230 | 1380790_at | - | - |
Rat Expression Set 230 | 1383723_PM_at | - | - |
Rat Expression Set 230 | 1383723_at | - | - |
Rat Expression Set 230 | 1380790_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10832584 | - | - |
Rat Genome U34 Set | rc_AI007963_at | - | - |
Rat Genome U34 Set | rc_AI180247_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22311685 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351400 | - | - |
