Assay Details
| Gene Symbol: Tmprss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107619.1 | NP_001101089.1 | 8 - 9 | 1008 | 79 | |
| XM_006256210.2 | XP_006256272.1 | 9 - 10 | 1098 | 79 | ||
| XM_006256211.2 | XP_006256273.1 | 9 - 10 | 1101 | 79 | ||
| XM_008772817.1 | XP_008771039.1 | 8 - 9 | 1490 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss3
Genomic Map