Assay Details
| Gene Symbol: RT1-Db1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008884.2 | NP_001008884.1 | 4 - 5 | 828 | 96 | |
| GenBank mRNA | AF084933.1 | - | 4 - 5 | 768 | 96 | |
| BC087691.1 | - | 5 - 6 | 748 | 7310949 | 96 | |
| CV111238.1 | - | 2 - 3 | 96 | |||
| FQ225193.1 | - | 4 - 5 | 816 | 96 | ||
| FQ226724.1 | - | 4 - 5 | 816 | 96 | ||
| FQ227087.1 | - | 4 - 5 | 813 | 96 | ||
| FQ228005.1 | - | 4 - 5 | 816 | 96 | ||
| FQ228129.1 | - | 4 - 5 | 816 | 96 | ||
| FQ228185.1 | - | 4 - 5 | 834 | 96 | ||
| FQ230360.1 | - | 4 - 5 | 816 | 96 | ||
| FQ231387.1 | - | 4 - 5 | 816 | 96 | ||
| FQ231709.1 | - | 4 - 5 | 816 | 96 | ||
| FQ232046.1 | - | 4 - 5 | 816 | 96 | ||
| FQ232100.1 | - | 4 - 5 | 815 | 96 | ||
| FQ232453.1 | - | 4 - 5 | 816 | 96 | ||
| FQ233049.1 | - | 4 - 5 | 812 | 96 | ||
| FQ233335.1 | - | 4 - 5 | 816 | 96 | ||
| FQ233396.1 | - | 4 - 5 | 816 | 96 | ||
| FQ233493.1 | - | 4 - 5 | 816 | 96 | ||
| FQ233800.1 | - | 4 - 5 | 794 | 96 | ||
| FQ233801.1 | - | 4 - 5 | 818 | 96 | ||
| FQ234464.1 | - | 4 - 5 | 815 | 96 | ||
| FQ234907.1 | - | 4 - 5 | 816 | 96 | ||
More Information
Set Membership: |
|

RT1-Db1
Genomic Map