Assay Details
| Gene Symbol: Hmga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256161.1 | XP_006256223.1 | 2 - 3 | 255 | 95 | |
| XM_006256162.2 | XP_006256224.1 | 2 - 3 | 291 | 95 | ||
| XM_006256163.2 | XP_006256225.1 | 2 - 3 | 666 | 95 | ||
| GenBank mRNA | AF511040.1 | - | 1 - 2 | 128 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hmga1
Genomic Map