Assay Details
| Gene Symbol: Papola | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108056.2 | NP_001101526.1 | 16 - 17 | 1730 | 72 | |
| XM_006240521.2 | XP_006240583.1 | 16 - 17 | 1740 | 72 | ||
| XM_008764807.1 | XP_008763029.1 | 16 - 17 | 1740 | 72 | ||
| XM_008764808.1 | XP_008763030.1 | 16 - 17 | 1740 | 72 | ||
| XM_008764809.1 | XP_008763031.1 | 16 - 17 | 1740 | 72 | ||
| XM_008764810.1 | XP_008763032.1 | 16 - 17 | 1739 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Papola
Genomic Map