Assay Details
Gene Symbol: Klc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081972.1 | NP_001075441.1 | 11 - 12 | 1489 | 74 | |
NM_001081973.1 | NP_001075442.1 | 11 - 12 | 1512 | 74 | ||
NM_001081974.1 | NP_001075443.1 | 11 - 12 | 1498 | 74 | ||
XM_006240621.2 | XP_006240683.1 | 11 - 12 | 2082 | 74 | ||
XM_006240622.2 | XP_006240684.1 | 11 - 12 | 2097 | 74 | ||
XM_006240623.2 | XP_006240685.1 | 11 - 12 | 2083 | 74 | ||
XM_006240625.2 | XP_006240687.1 | 11 - 12 | 1978 | 74 | ||
XM_006240626.2 | XP_006240688.1 | 11 - 12 | 1977 | 74 | ||
XM_006240627.2 | XP_006240689.1 | 11 - 12 | 1978 | 74 | ||
XM_006240628.2 | XP_006240690.1 | 11 - 12 | 1978 | 74 | ||
XM_006240629.2 | XP_006240691.1 | 11 - 12 | 1977 | 74 | ||
XM_006240630.2 | XP_006240692.1 | 11 - 12 | 1978 | 74 | ||
GenBank mRNA | M75146.1 | - | 11 - 12 | 1489 | 74 | |
M75147.1 | - | 11 - 12 | 1512 | 74 | ||
M75148.1 | - | 11 - 12 | 1498 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
