Assay Details
Gene Symbol: Adssl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750237.3 | XP_003750285.1 | 8 - 9 | 874 | 59 | |
XM_006240653.2 | XP_006240715.1 | 8 - 9 | 867 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
