Assay Details
| Gene Symbol: NEWGENE_1565481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773047.1 | XP_008771269.1 | 3 - 4 | 725 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1565481| Gene Symbol: NEWGENE_1565481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773047.1 | XP_008771269.1 | 3 - 4 | 725 | 134 | |
Set Membership: |
|