Assay Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033653.1 | NP_387502.1 | 11 - 12 | 1167 | 69 | |
| GenBank mRNA | BC166508.1 | - | 11 - 12 | 1150 | 69 | |
| D00688.1 | - | 11 - 12 | 1173 | 69 | ||
| FM031063.1 | - | 4 - 5 | 251 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Maoa
Genomic Map