Assay Details
| Gene Symbol: RGD621098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139040.1 | NP_620609.1 | 11 - 12 | 1172 | 71 | |
| GenBank mRNA | U92010.1 | - | 11 - 12 | 1172 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD621098
Genomic Map