Assay Details
| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012165.1 | NP_001012165.1 | 1 - 2 | 329 | 63 | |
| GenBank mRNA | BC089914.1 | - | 1 - 2 | 329 | 7305045 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Emc8
Genomic Map