Assay Details
| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012165.1 | NP_001012165.1 | 3 - 4 | 485 | 71 | |
| GenBank mRNA | BC089914.1 | - | 3 - 4 | 485 | 7305045 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Emc8
Genomic Map