Assay Details
| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005381.2 | NP_001005381.2 | 2 - 3 | 183 | 119 | |
| GenBank mRNA | BC079094.1 | - | 2 - 3 | 154 | 7123989 | 119 |
| CB582878.1 | - | 2 - 3 | 183 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P480636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158843 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P480636 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P480636 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P480636 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P480636 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64282 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63219 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42859 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90975 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50654 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10811042 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811047 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811049 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811051 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811045 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811052 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811050 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811046 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811044 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811043 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811048 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811055 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811057 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10811041 | - | - |
| Rat Genome 230 2.0 Array | 1378934_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383559_at | - | - |
| Rat Genome 230 2.0 Array | 1381134_at | - | - |
| Rat Genome 230 2.0 Array | 1381134_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1396661_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388814_at | - | - |
| Rat Genome 230 2.0 Array | 1383559_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388814_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1396661_at | - | - |
| Rat Genome 230 2.0 Array | 1378934_at | - | - |
| Affymetrix HT Rat Focus Array | 1388814_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388814_at | - | - |
| Affymetrix HT Rat Focus Array | 1396661_at | - | - |
| Affymetrix HT Rat Focus Array | 1396661_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900005155.rn.1 | TC1900000604.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900005160.rn.1 | TC1900000604.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900005156.rn.1 | TC1900000604.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900005158.rn.1 | TC1900000604.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900005161.rn.1 | TC1900000604.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388814_PM_at | - | - |
| Rat Expression Set 230 | 1388814_at | - | - |
| Rat Expression Set 230 | 1396661_at | - | - |
| Rat Expression Set 230 | 1383559_at | - | - |
| Rat Expression Set 230 | 1396661_PM_at | - | - |
| Rat Expression Set 230 | 1383559_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10811041 | - | - |
| Rat Genome U34 Set | rc_AI045477_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21304845 | - | - |

Ddx19a
Genomic Map