Assay Details
Gene Symbol: Wwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106184.1 | NP_001099654.1 | 3 - 4 | 781 | 58 | |
GenBank mRNA | BC168152.1 | - | 3 - 4 | 750 | 7461024 | 58 |
JN712752.1 | - | 793 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
