Assay Details
| Gene Symbol: Cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017202.1 | NP_058898.1 | 2 - 3 | 162 | 106 | |
| GenBank mRNA | BC084719.1 | - | 1 - 2 | 78 | 7317602 | 106 |
| FQ211185.1 | - | 2 - 3 | 115 | 106 | ||
| FQ214265.1 | - | 2 - 3 | 171 | 106 | ||
| FQ217878.1 | - | 2 - 3 | 130 | 106 | ||
| FQ223981.1 | - | 2 - 3 | 124 | 106 | ||
| FQ227128.1 | - | 2 - 3 | 160 | 106 | ||
| FQ228805.1 | - | 2 - 3 | 112 | 105 | ||
| X14209.1 | - | 2 - 3 | 144 | 106 | ||
| X15029.1 | - | 1 - 2 | 89 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox4i1
Genomic Map