Assay Details
| Gene Symbol: Anapc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108445.1 | NP_001101915.1 | 2 - 3 | 279 | 88 | |
| XM_006255406.2 | XP_006255468.1 | 2 - 3 | 281 | 88 | ||
| XM_008772516.1 | XP_008770738.1 | 2 - 3 | 235 | 88 | ||
| GenBank mRNA | BC168746.1 | - | 2 - 3 | 201 | 6920404 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc10
Genomic Map