Assay Details
Gene Symbol: Crispld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138518.2 | NP_612527.2 | 12 - 13 | 1373 | 57 | |
XM_006255708.2 | XP_006255770.1 | 13 - 14 | 1496 | 57 | ||
GenBank mRNA | AF109674.1 | - | 12 - 13 | 1387 | 57 | |
CK367026.1 | - | 6 - 7 | 437 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
