Assay Details
Gene Symbol: RGD1304884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107436.1 | NP_001100906.1 | 12 - 13 | 1456 | 83 | |
XM_006255743.2 | XP_006255805.1 | 10 - 11 | 1274 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P117413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156786 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117413 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117413 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P117413 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P372950 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10808401 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808399 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808396 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808398 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808406 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808397 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808400 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808395 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808405 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808403 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808402 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808404 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10808393 | - | - |
Rat Genome 230 2.0 Array | 1382737_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376583_at | - | - |
Rat Genome 230 2.0 Array | 1382737_at | - | - |
Rat Genome 230 2.0 Array | 1376583_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382737_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382737_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900006052.rn.1 | TC1900000726.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900006053.rn.1 | TC1900000726.rn.1 | RTA TC |
Rat Expression Set 230 | 1376583_at | - | - |
Rat Expression Set 230 | 1376583_PM_at | - | - |
Rat Expression Set 230 | 1382737_PM_at | - | - |
Rat Expression Set 230 | 1382737_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808393 | - | - |
Rat Genome U34 Set | rc_AI059118_at | - | - |
Rat Genome U34 Set | rc_AI227724_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22117285 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357555 | - | - |
