Assay Details
| Gene Symbol: Sf3b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106187.2 | NP_001099657.2 | 10 - 11 | 1498 | 65 | |
| XM_006255620.2 | XP_006255682.1 | 10 - 11 | 1633 | 65 | ||
| GenBank mRNA | BC168883.1 | - | 4 - 5 | 403 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sf3b3
Genomic Map