Assay Details
| Gene Symbol: RGD1309651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_341702.5 | XP_341703.1 | 3 - 4 | 1050 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309651| Gene Symbol: RGD1309651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_341702.5 | XP_341703.1 | 3 - 4 | 1050 | 124 | |
Set Membership: |
|