Assay Details
| Gene Symbol: Asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100505.1 | NP_001093975.1 | 3 - 4 | 463 | 63 | |
| XM_008772367.1 | XP_008770589.1 | 4 - 5 | 1284 | 63 | ||
| XM_008772368.1 | XP_008770590.1 | 4 - 5 | 1412 | 63 | ||
| XM_008772369.1 | XP_008770591.1 | 4 - 5 | 1407 | 63 | ||
| GenBank mRNA | BC098819.1 | - | 3 - 4 | 398 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asna1
Genomic Map