Assay Details
Gene Symbol: Kiaa0895l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044292.1 | NP_001037757.1 | 4 - 5 | 1336 | 74 | |
XM_008772533.1 | XP_008770755.1 | 4 - 5 | 1994 | 74 | ||
GenBank mRNA | BC098045.1 | - | 3 - 4 | 907 | 7457085 | 74 |
BC098942.1 | - | 4 - 5 | 1336 | 7456844 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
