Assay Details
| Gene Symbol: Prmt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191599.1 | NP_001178528.1 | 4 - 5 | 866 | 90 | |
| XM_006255424.2 | XP_006255486.1 | 4 - 5 | 902 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prmt10
Genomic Map