Assay Details
| Gene Symbol: Asf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107160.1 | NP_001100630.1 | 2 - 3 | 392 | 119 | |
| GenBank mRNA | BC158705.1 | - | 2 - 3 | 318 | 9030040 | 119 |
More Information
Updated Mapping Information
Set Membership: |
|

Asf1b
Genomic Map