Assay Details
| Gene Symbol: Wdr59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772573.1 | XP_008770795.1 | 11 - 12 | 1126 | 60 | |
| XM_008772574.1 | XP_008770796.1 | 11 - 12 | 1126 | 60 | ||
| XM_008772575.1 | XP_008770797.1 | 11 - 12 | 1126 | 60 | ||
| XM_008772576.1 | XP_008770798.1 | 11 - 12 | 1127 | 60 | ||
| XM_008772577.1 | XP_008770799.1 | 11 - 12 | 1127 | 60 | ||
| XM_008772578.1 | XP_008770800.1 | 11 - 12 | 1125 | 60 | ||
| XM_008772579.1 | XP_008770801.1 | 11 - 12 | 1126 | 60 | ||
| XM_008772580.1 | XP_008770802.1 | 11 - 12 | 1126 | 60 | ||
| XM_008772581.1 | XP_008770803.1 | 11 - 12 | 1126 | 60 | ||
| XM_008772582.1 | XP_008770804.1 | 11 - 12 | 1126 | 60 | ||
More Information
Set Membership: |
|

Wdr59
Genomic Map