Assay Details
| Gene Symbol: St3gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255625.2 | XP_006255687.1 | 8 | 113 | ||
| XM_006255626.2 | XP_006255688.1 | 9 | 113 | |||
| XM_006255627.2 | XP_006255689.1 | 7 | 113 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

St3gal2
Genomic Map