Assay Details
| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106182.2 | NP_001099652.1 | 3 - 4 | 1445 | 87 | |
| GenBank mRNA | BC168966.1 | - | 3 - 4 | 1448 | 7124140 | 87 |
More Information
Updated Mapping Information
Set Membership: |
|

Cog8
Genomic Map