Assay Details
| Gene Symbol: Tradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255442.2 | XP_006255504.1 | 1 - 2 | 68 | 64 | |
| XM_006255444.2 | XP_006255506.1 | 1 - 2 | 67 | 64 | ||
| GenBank mRNA | FQ215613.1 | - | 1 - 2 | 41 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006255442.2, XM_006255444.2 transcripts.
Set Membership: |
|

Tradd
Genomic Map