Assay Details
| Gene Symbol: NEWGENE_1305243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_341690.7 | XP_341691.7 | 2247 | 60 | ||
| GenBank mRNA | FQ230829.1 | - | 2199 | 60 | ||
| Gene Symbol: Rfwd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255619.2 | XP_006255681.1 | 4 - 5 | 779 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NEWGENE_1305243
Genomic Map