Assay Details
Gene Symbol: Enkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106180.1 | NP_001099650.1 | 2 - 3 | 563 | 121 | |
XM_006255476.2 | XP_006255538.1 | 3 - 4 | 343 | 121 | ||
XM_006255477.2 | XP_006255539.1 | 3 - 4 | 393 | 121 | ||
XM_006255478.2 | XP_006255540.1 | 3 - 4 | 327 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
