Assay Details
Gene Symbol: Mlkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772570.1 | XP_008770792.1 | 11 - 12 | 1712 | 58 | |
XM_008772571.1 | XP_008770793.1 | 11 - 12 | 1667 | 58 | ||
XM_008772572.1 | XP_008770794.1 | 10 - 11 | 1578 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
