Assay Details
| Gene Symbol: Clec18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255608.2 | XP_006255670.1 | 4 - 5 | 1289 | 63 | |
| XM_006255609.2 | XP_006255671.1 | 5 - 6 | 1175 | 63 | ||
| XM_006255610.2 | XP_006255672.1 | 4 - 5 | 1289 | 63 | ||
| XM_006255611.2 | XP_006255673.1 | 4 - 5 | 1289 | 63 | ||
| XM_006255612.2 | XP_006255674.1 | 4 - 5 | 1289 | 63 | ||
| XM_008772568.1 | XP_008770790.1 | 4 - 5 | 1289 | 63 | ||
More Information
Set Membership: |
|

Clec18a
Genomic Map