Assay Details
| Gene Symbol: Dhodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008553.1 | NP_001008553.1 | 3 - 4 | 486 | 78 | |
| XM_006255601.2 | XP_006255663.1 | 3 - 4 | 659 | 78 | ||
| XM_008772529.1 | XP_008770751.1 | 3 - 4 | 659 | 78 | ||
| GenBank mRNA | X80778.1 | - | 3 - 4 | 486 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhodh
Genomic Map