Assay Details
| Gene Symbol: Ptger1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278475.1 | NP_001265404.1 | 1 - 2 | 282 | 106 | |
| XM_008772362.1 | XP_008770584.1 | 1 - 2 | 934 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278475.1 transcripts.
Set Membership: |
|

Ptger1
Genomic Map