Assay Details
Gene Symbol: Aprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013061.1 | NP_001013079.1 | 3 - 4 | 323 | 94 | |
GenBank mRNA | FQ218973.1 | - | 350 | 94 | ||
FQ228571.1 | - | 3 - 4 | 341 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
