Assay Details
| Gene Symbol: Chmp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083313.1 | NP_001076782.1 | 3 - 4 | 179 | 80 | |
| GenBank mRNA | CK358527.1 | - | 3 - 4 | 158 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chmp1a
Genomic Map