Assay Details
| Gene Symbol: LOC100360750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728002.4 | XP_002728048.1 | 1 | 268 | 99 | |
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126085.1 | NP_001119557.1 | 3 - 4 | 295 | 99 | |
| XM_006255408.1 | XP_006255470.1 | 2 - 3 | 210 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100360750
Genomic Map