Assay Details
Gene Symbol: Pskh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108897.1 | NP_001102367.1 | 1 - 2 | 115 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108897.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P112846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109870 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P112846 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P112846 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P112846 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19053 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14252 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103257 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94761 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10807431 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10807434 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10807430 | - | - |
Rat Genome 230 2.0 Array | 1390288_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390288_at | - | - |
Affymetrix HT Rat Focus Array | 1390288_at | - | - |
Affymetrix HT Rat Focus Array | 1390288_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900004297.rn.1 | TC1900000497.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900004291.rn.1 | TC1900000497.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900004296.rn.1 | TC1900000497.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900004292.rn.1 | TC1900000497.rn.1 | RTA TC |
Rat Expression Set 230 | 1390288_at | - | - |
Rat Expression Set 230 | 1390288_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10807430 | - | - |
Rat Genome U34 Set | rc_AI013362_at | - | - |
Rat Genome U34 Set | rc_AI058755_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22149648 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373510 | - | - |
