Assay Details
| Gene Symbol: Ucp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012682.2 | NP_036814.1 | 1 - 2 | 307 | 90 | |
| GenBank mRNA | BC088156.1 | - | 1 - 2 | 295 | 7380759 | 90 |
| M11814.1 | - | 1 - 2 | 305 | 90 | ||
| X03894.1 | - | 1 - 2 | 217 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ucp1
Genomic Map