Assay Details
| Gene Symbol: Mlycd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053477.1 | NP_445929.1 | 3 - 4 | 789 | 98 | |
| GenBank mRNA | AF304865.1 | - | 3 - 4 | 789 | 98 | |
| AJ007704.1 | - | 3 - 4 | 789 | 98 | ||
| BC061845.1 | - | 3 - 4 | 792 | 5621015 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlycd
Genomic Map