Assay Details
| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053866.3 | NP_446318.3 | 10 - 11 | 1602 | 62 | |
| GenBank mRNA | CV110320.1 | - | 3 - 4 | 462 | 62 | |
| FQ211509.1 | - | 1663 | 62 | |||
| U17901.1 | - | 10 - 11 | 1461 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plaa
Genomic Map