Assay Details
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033709.1 | NP_001028881.1 | 11 - 12 | 1254 | 114 | |
GenBank mRNA | BC101850.1 | - | 11 - 12 | 1254 | 5621707 | 114 |
FQ210318.1 | - | 11 - 12 | 1269 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P107923 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P715449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300021 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108718 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P715449 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P107923 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P715449 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P107923 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P107923 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P715449 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P107923 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52273 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86341 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37706 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42012 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33888 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10870149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870155 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870162 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870159 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870158 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870157 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870161 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870160 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10870146 | - | - |
Affymetrix Rat Clariom S Assay | PSR0500006678.rn.1 | TC0500001048.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10870146 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21708604 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362432 | - | - |
