Assay Details
| Gene Symbol: Ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017135.3 | NP_058831.1 | 2 - 3 | 263 | 88 | |
| XM_006238445.2 | XP_006238507.1 | 3 - 4 | 568 | 88 | ||
| GenBank mRNA | BC087024.1 | - | 2 - 3 | 315 | 7107348 | 88 |
| D87809.1 | - | 2 - 3 | 263 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ak4
Genomic Map