Assay Details
| Gene Symbol: Pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256432.2 | XP_006256494.1 | 1 - 2 | 369 | 84 | |
| XM_346131.7 | XP_346132.5 | 1 - 2 | 368 | 84 | ||
More Information
Set Membership: |
|

Pla2g12b
Genomic Map