Assay Details
| Gene Symbol: Pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256432.2 | XP_006256494.1 | 3 - 4 | 617 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g12b| Gene Symbol: Pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256432.2 | XP_006256494.1 | 3 - 4 | 617 | 76 | |
Set Membership: |
|